Precoiled Pipe Markers- Direction Arrow White/Blue, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
A4118 B4118 C4118 D4118 E4118 F4118 G4118 H4118
94003-516EA 28.11 CAD
94003-516 94004-290 94005-760 94009-704 94012-778 94013-974 94014-520 94015-300
Precoiled Pipe Markers- Direction Arrow White/Blue, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007