Precoiled Pipe Markers- BLANK RED, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
A6001 B6001
94003-522EA 28.11 CAD
94003-522 94004-296
Precoiled Pipe Markers- BLANK RED, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007.