Precoiled Pipe Markers 'Oil', Black/Yellow, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
A4077 B4077 C4077 D4077 E4077 F4077 G4077 H4077
94003-434EA 28.11 CAD
94003-434 94004-208 94005-648 94009-580 94012-696 94013-892 94014-438 94015-218
Precoiled Pipe Markers 'Oil', Black/Yellow, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007.