Precoiled Pipe Markers 'Natural Gas', Black/Yellow, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
A4073 B4073 C4073 D4073 E4073 F4073 G4073 H4073
94003-426EA 28.11 CAD
94003-426 94004-200 94005-640 94009-572 94012-688 94013-884 94014-430 94015-210
Precoiled Pipe Markers 'Natural Gas', Black/Yellow, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007.