The Defender Detectable Warning Tape, NMC (National Marker Company)C
Supplier: ACCUFORM MANUFACTURING, INC
DT2 BIRR DT2 BNPW DT2 BPW DT2 BRFM DT2 BW DT2 GS DT2 GSD DT2 OCATV DT2 OFBO DT2 OTC DT2 PRW DT2 RE DT2 YG DT3 BIRR DT3 BNPW DT3 BPW DT3 BRFM DT3 BW DT3 GS DT3 GSD DT3 OCATV DT3 OFBO DT3 OTC DT3 PRW DT3 RE DT3 YG DT6 BNPW DT6 BPW DT6 BRFM DT6 BW DT6 GS DT6 GSD DT6 OCATV DT6 OFBO DT6 OTC DT6 PRW DT6 RE DT6 YG
94012-468EA 322.46 CAD
94012-468 94012-470 94012-472 94012-474 94012-476 94012-478 94012-480 94012-482 94012-484 94012-486 94012-488 94012-490 94012-492 94012-494 94012-496 94012-498 94012-500 94012-502 94012-504 94012-506 94012-508 94012-510 94012-512 94012-514 94012-516 94012-518 94012-520 94012-522 94012-524 94012-526 94012-528 94012-530 94012-532 94012-534 94012-536 94012-538 94012-540 94012-542
The Defender Detectable Warning Tape, NMC (National Marker Company)C
Tapes
Minimize field costs and time spent locating buried lines
Made from 5 mil., triple layer lamination of aluminum foil core encased in 100% virgin polyethylene, APWA (American Public Works Association) color coded, legend specific. Withstands temperatures from - 94⁰F to 200⁰F for longevity. Subsurface graphics permanently protected from soil acids and alkalis.