Precoiled Pipe Markers- 'Nitrogen', White/Green, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
A4074 B4074 C4074 D4074 E4074 F4074 G4074 H4074
94003-428EA 27.98 CAD
94003-428 94004-202 94005-642 94009-574 94012-690 94013-886 94014-432 94015-212
Precoiled Pipe Markers- 'Nitrogen', White/Green, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007