Precoiled Pipe Markers- 'Cooling Water Supply', White/Green, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
A4119 B4119 C4119 D4119 E4119 F4119 G4119 H4119
94003-518EA 28.11 CAD
94003-518 94004-292 94005-762 94009-706 94012-780 94013-976 94014-522 94015-302
Precoiled Pipe Markers- 'Cooling Water Supply', White/Green, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas
Certifications: Meets ANSI A13.1-2007