Precoiled Pipe Markers- 'Sprinkler Fire' W/R, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
A4095 B4095 C4095 D4095 E4095 F4095 G4095 H4095
94003-470EA 28.11 CAD
94003-470 94004-244 94005-702 94009-632 94012-732 94013-928 94014-474 94015-254
Precoiled Pipe Markers- 'Sprinkler Fire' W/R, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007