Robotic Tips for Agilent Automated Platforms, Biotix
Supplier: Biotix
63300093 63300094
89202-238CS 7129.18 CAD
89202-238 89129-584
Robotic Tips for Agilent Automated Platforms, Biotix
Pipette Tips
Biotix robotic tips for Agilent™ VPrep® and Bravo® platforms are engineered to ensure complete compatibility, requiring no changes in programming scripts in the platform's liquid handling software suite.
All Biotix robotic products are manufactured to the highest engineering standards for robotic tips. For this reason, no adjustments to the instrument should be necessary to use Biotix robotic tips. The standard tip definitions bundled in the robotic software program will suffice.
These tips feature Statix™, a proprietary technology that removes static and prevents automation failure; Blade™ technology, which eliminates hanging droplets and the need for tip touch-off with even the most challenging reagents; and X-Resin™, an exclusive polypropylene resin for low sample retention.
Manufactured to the highest engineering standards for robotic systems, these ultra-straight tips target the center of microplate wells precisely, delivering exceptional accuracy and precision without the risk of jamming. Made of medical-grade virgin polypropylene, these tips are BioReady™ certified to be free of RNase, DNase, and Endotoxins (pyrogens). They are available in standard or filtered, nonsterile or pre-sterile formats.
Certifications: Certified free of RNase, DNase, pyrogens, and PCR Inhibitors. Manufactured under stringent controls in an ISO 9001 certified facility. Each and every lot is tested for accuracy and precision, ensuring superior quality and compatibility. All raw materials are controlled.
Ordering information: Biotix tips are assembled in racks that are SBS compliant.