Instatherm® Round Bottom Flasks with Three Necks, Ace Glass Incorporated
Supplier: Ace Glass
9642-127 9642-129 9642-131 9642-133 9642-135 9642-137
89075-420EA 1391.19 CAD
89075-420 89075-422 89075-424 76291-862 76291-864 76291-866
Instatherm® Round Bottom Flasks with Three Necks, Ace Glass Incorporated
Flasks Round Bottom Flasks
Distilling round-bottom flask coated with Ace Instatherm®.
Three-prong connector plug is designed to work with Ace temperature controllers where the output voltage can be automatically limited to match the flask. Flask can also be matched with ACE temperature thermocouple sensors for complete automatic operation.
Designed to operate up to 250 °C, heating response time is very rapid, (5° per minute), with very low thermal lag. Has 24/40 vertical standard taper reinforced outer center neck and two 24/40 angled standard taper reinforced outer side necks.
Delivery information: Comes complete with integral six-foot controller cord.
Caution: Caution: Never run Instatherm® flasks dry or breakage can occur, especially at higher temperatures.