Instatherm® Flask, Two Necks, Round Bottom, Ace Glass Incorporated
Supplier: Ace Glass
9637-136 9637-138 9637-170
89075-414EA 951.12 CAD
89075-414 89075-416 89075-418
Instatherm® Flask, Two Necks, Round Bottom, Ace Glass Incorporated
Flasks Round Bottom Flasks
Distilling round-bottom flask coated with Ace Instatherm®
Three-prong connector plug is designed to work with Ace temperature controllers where the output voltage can be automatically limited to match the flask. Flask can also be matched with ACE temperature thermocouple sensors for complete automatic operation.
Caution: Never run Instatherm® flasks dry or breakage can occur, especially at higher temperatures.
Designed to operate up to 250ºC, heating response time is very rapid, (5° per minute), with very low thermal lag. Has vertical standard taper reinforced outer center neck and angled standard taper reinforced outer side neck.
Ordering information: Comes complete with integral six-foot controller cord.