Instatherm® Six-Foot Controller Cords, Ace Glass Incorporated
Supplier: Ace Glass
9698-05 9698-10 9698-16 9698-20
89075-440EA 122.9 CAD
89075-440 89075-442 89075-444 71000-042
Instatherm® Six-Foot Controller Cords, Ace Glass Incorporated
Controllers Temperature Controllers
Controller cords with various connectors for connecting all Ace Instatherm® vessels to temperature controllers.
Ordering information: Two versions (89075-444 and 71000-042) have "fast-acting" fuse to protect both the vessel and the controller from over-voltage or surges.