Instatherm® Flask, Single Neck, Round Bottom, Ace Glass Incorporated
Supplier: Ace Glass
9635-158 9635-160
89075-410EA 1259.04 CAD
89075-410 89075-412
Instatherm® Flask, Single Neck, Round Bottom, Ace Glass Incorporated
Flasks Round Bottom Flasks
Distilling, round-bottom flask with Instatherm® provide a convenient, safe heating of contents
Caution: Never run Instatherm® baths dry or breakage can occur, especially at higher temperatures.
Reinforced standatd taper 24/40 outer joint at top. Designed to operate up to 250ºC, heating response time is very rapid, (5° per minute), with very low thermal lag. Three-prong connector plug is designed to work with Ace temperature controllers where the output voltage can be automatically limited to match the bath. Flask can also be matched with ACE temperature thermocouple sensors for complete automatic operation. If using another type temperature controller, voltage should not exceed the rated voltage (stated below). Integral six-foot controller cord.