Instatherm® Erlenmeyer Flask, Ace Glass Incorporated
Supplier: Ace Glass
Instatherm®
9633-08 9633-14 9633-10
71000-046EA 1283.09 CAD
71000-046 71000-044 71000-048
Instatherm® Erlenmeyer Flask, Ace Glass Incorporated
Flasks Erlenmeyer Flasks
Erlenmeyer flask with Instatherm® provide a convenient, safe heating of contents in the popular Erlenmeyer flask format for bench or magnetic stirrer use.
Designed to operate up to 250ºC, heating response time is very rapid, (5° per minute), with very low thermal lag. Three-prong connector plug is designed to work with Ace temperature controllers where the output voltage can be automatically limited to match the bath. Flask can also be matched with ACE temperature thermocouple sensors for complete automatic operation. If using another type temperature controller, voltage should not exceed the rated voltage (stated below). Integral six-foot controller cord.
Caution: Never run Instatherm® baths dry or breakage can occur, especially at higher temperatures.