Instatherm® Reaction Flask, Ace Glass Incorporated
Supplier: Ace Glass
9656-08 9656-12
89075-436EA 2670.51 CAD
89075-436 89075-438
Instatherm® Reaction Flask, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Flask Shape
Rugged cylindrical reaction flask and head coated with Ace Instatherm®
Caution: Never run Instatherm® flasks dry or breakage can occur, especially at higher temperatures.
Has flat, ground 4" conical flange. Designed to operate up to 250ºC, heating response time is very rapid, (5° per minute), with very low thermal lag. Three-prong connector plug is designed to work with Ace temperature controllers where the output voltage can be automatically limited to match the bath. Flask can also be matched with ACE temperature thermocouple sensors for complete automatic operation. Flask is coatedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable