Instatherm® Reaction Flask/Head, Ace Glass Incorporated
Supplier: Ace Glass
9655-17 9655-22 9655-33 9655-37 9655-39
89075-426EA 3037.8 CAD
89075-426 89075-428 89075-430 89075-432 89075-434
Instatherm® Reaction Flask/Head, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Flask Shape
Rugged cylindrical reaction flask and head coated with Ace Instatherm®
Flask is coated to within approximately 38 mm of the flange. Heads have vertical standard taper reinforced outer center neck and three vertical standard taper reinforced side necks, and require 89075-444 connecting cord (not supplied).
Caution: Never run Instatherm® flasks dry or breakage can occur, especially at higher temperatures.
Duran flat, ground 4" flange with o-ring groove for use with 89055-110 quick release clamp. Designed to operate up to 250°C, heating response time is very rapid, (5° per minute), with very low thermal lag. Three-prong connector plug is designed to work with Ace temperature controllers where the output voltage can be automatically limited to match the bath. Flask can also be matched with ACE temperature thermocouple sensors for complete automatic operation.
Ordering information: Flask is supplied with CAPFE (PTFE encapsulated silicone) o-ring.