Finntip Stepper Tips, Thermo Scientific
Supplier: Thermo Fisher Scientific
9404203 9404200 9404233 9404223 9404213 9404193 9404183 9404173 9404230 9404220 9404210 9404190 9404180 9404170
14222-046PK 122.28 CAD
14222-046 53515-226 53515-916 53515-918 53515-920 53515-924 53515-926 53515-928 53515-930 53515-932 53515-934 53515-936 53515-938 53515-940
Finntip Stepper Tips, Thermo Scientific
Pipette Tips
Finntip stepper tips are the perfect fit with Thermo Scientific Finnpipette stepper pipetters.
The finntip stepper tips are plunger-style tips for positive displacement pipetting. Seven sizes are available in sterile and non sterile versions.