Wiper Dispensers, S-Curve Technologies
Supplier: S-Curve Technologies
WDL-99CF WDL-99CF P WDL-125CF WDL-125CF P WDL-39CF WDL-39CF P WDL-312CF WDL-312CF P
89208-594EA 424.1 CAD
89208-594 89208-596 89208-598 89208-600 89208-602 89208-604 89208-606 89208-608
Wiper Dispensers, S-Curve Technologies
Wipe Dispensers
Dispensers feature a closed front for 22.9x22.9cm (9x9") and 30.5x30.5cm (12x12") wipers
Fully enclosed dispenser for wipers with full-length hinged lid for durability. Dispensers hold either large or medium quantity of wipers. Wall mounted or free standing.
Ordering information: Wipers sold separately.
Order Now
SPECIFICATIONS
Environmentally Preferable