Finntip™ Multistepper Pipette Tips, Thermo Scientific
Supplier: Thermo Fisher Scientific
94052450 9401300 9401330
11310-847CS 607.24 CAD
11310-847 53516-014 53516-016
Finntip™ Multistepper Pipette Tips, Thermo Scientific
Pipette Tips
Finntip Multistepper Pipette Tips optimize the performance of Thermo Scientific Finnpipette multistepper pipetters.
  • Accommodates standard 9 mm microplate spacing
  • Sterile or non-sterile versions
They are specifically designed for use with the Thermo Scientific Finnpipette Multistepper Pipetter. This tip has a nominal volume of 1500 μl and accommodates standard 9 mm microplate spacing. Finntip multistepper tips are available in racks of 96 tips, in both sterile and non-sterile versions.
Order Now
SPECIFICATIONS
Environmentally Preferable