Matrix Filtered D.A.R.T.'s Tips, Thermo Scientific
Supplier: Thermo Fisher Scientific
5318 5328 5418 5508 5538 5548 5588
CA89168-924CS 12810.92 CAD
CA89168-924 CA89168-930 CA89168-936 CA89168-942 CA89168-960 CA89168-966 CA89168-972
Matrix Filtered D.A.R.T.'s Tips, Thermo Scientific
Pipette Tips
Unlike conventional automation tips, Matrix D.A.R.T.s tips are provided in disposable, rigid magazines that bond onto the pipetting heads by sealing against a silicone pad.
Compatible with the Versette, Hydra DT, PlateMate 2x3 and PlateMate Plus Automated Pipetting Systems. Stepped-body design minimizes volume displacement and provides tip straightness for better well access.
This forms a definitive seal that minimizes tip-to-tip height variations and eliminates static interactions, delivering.