Matrix D.A.R.T.'s Tips, Thermo Scientific
Supplier: Thermo Fisher Scientific
5316 5317 5327 5416 5417 5506-11 5517-11 5518-11 5526 5527 5536 5537 5547 5586 5587
CA89168-920CS 8952.34 CAD
CA89168-920 CA89168-922 CA89168-928 CA89168-932 CA89168-934 CA89168-938 CA89168-946 CA89168-948 CA89168-950 CA89168-952 CA89168-956 CA89168-958 CA89168-964 CA89168-968 CA89168-970
Matrix D.A.R.T.'s Tips, Thermo Scientific
Pipette Tips
Unlike conventional automation tips, Matrix D.A.R.T.s tips are provided in disposable, rigid magazines that bond onto the pipetting heads by sealing against a silicone pad.
  • Disposable, rigid magazines
  • Definitive seal minimizes tip-to-tip height variations
  • Consistent dispensing into the bottom of microplate wells
Compatible with the Versette, Hydra DT, PlateMate 2×3 and PlateMate Plus Automated Pipetting Systems. Stepped-body design minimizes volume displacement and provides tip straightness for better well access. Disposable, rigid magazines form aMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable