Pipette Tips for AP96, FX/NX, 1000, 2000, and 3000 Series Workstations, Thermo Scientific
Supplier: Thermo Scientific
141-96RS 142-96R 142-96RS 912-262 912-021 915-261 915-262 917-261 917-262 919-021G 935-021 918-011 919-252 918-261-05 919-021-05 916-021 916-261 916-262 918-252 919-011
46300-360PK 32.62 CAD
46300-360 46300-362 46300-370 46300-372 53226-016 53226-018 72830-210 72830-212 72830-216 72830-218 72830-290 72830-353 72830-408 72830-412 72830-518 72830-528 89025-876 89025-878 89025-880 490014-540 76457-542
Pipette Tips for AP96, FX/NX, 1000, 2000, and 3000 Series Workstations, Thermo Scientific
Pipette Tips
Molecular BioProducts pipette tips are designed to fit AP96, FX/NX, 1000, 2000, and 3000 liquid handling workstations.
  • Designed for compatibility and performance
  • Variety of sizes and styles available
  • Lot-tested using 15-point program
BioRobotix® robotic pipette tips are available in low retention for critical samples and in extended length for increased volume and the ability to reach into deep well plates.
Genomic tips are excellent for pipetting of more viscous samples. The tips have an orifice that is up to 260% larger than the standardMore Product Information