Disposable Automation Reservoirs
Supplier: Avantor
1064-15-6 1064-05-7
89200-078CS 374.6 CAD
89200-078 89200-082 89200-084 89200-086 89200-088
Disposable Automation Reservoirs
Reagent Reservoirs
Disposable automation reservoirs are of polypropylene construction and are designed for simple, economical handling of reagents on automated systems.
The standard microplate dimensions enables reservoirs to be used in plate handlers or stackers.
With the same dimensions as standard microplates, these disposable reservoirs can be used in plate handlers or stackers. The removable splash baffles prevent reagents from splashing out when being moved on robotic decks. They are compatible with virtually any automated liquid handler as well as all of our automated pipette tips.