Precoiled Pipe Markers- 'Carbon Dioxide' B/Y, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
A4011 B4011 C4011 D4011 E4011 F4011 G4011 H4011
94003-302EA 28.11 CAD
94003-302 94004-076 94005-462 94009-376 94012-564 94013-760 94014-306 94015-086
Precoiled Pipe Markers- 'Carbon Dioxide' B/Y, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007