Microsep Advance Centrifugal Devices, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
MCP003C46 MCP030C46 MCPM02C68 MCP003C46 MCP010C41 MCP010C46 MCP030C41 MCP030C46 MCP100C41 MCP100C46 MCPM45C67 MCP003C41 MCPM02C67 MCPM45C67 MCPM45C68 MCP001C41 MCP001C46 MCPM02C68
89132-006PK 920.11 CAD
89132-006 89132-014 89132-022 CA89132-006 CA89132-008 CA89132-010 CA89132-012 CA89132-014 CA89132-016 CA89132-018 CA89132-024 CA89132-004 CA89132-020 89132-024 89132-026 CA89233-876 CA89233-878 CA89132-022
Microsep Advance Centrifugal Devices, Cytiva (Formerly Pall Lab)
Filters Centrifugal Filters
Precise, quick recovery of microliter volumes of concentrate from starting volumes up to 5.0 ml.
  • Available with low protein-binding OmegaTM and Supor® (PES) membranes
  • Omega modified PES membranes come in a wide range of MWCOs, color-coded for easy identification
  • With Omega membranes you can Achieve 50X concentration and >90% recovery in just minutes
  • Supor (PES) membranes come in 0.45 and 0.2 µm pore sizes for microfiltration applications
  • Constructed of low-binding polypropylene
  • Ultrasonically welded seals prevent bypass or seal failure
For smaller samples use Pall Nanosep ®centrifuge filters. For larger samples use Pall Macrosep® or Jumbosep™ centrifugal filter.
Microsep® Centrifugal devices with Omega (modified polyethersulfone) membrane for ultrafiltration are ideal for concentration of dilute protein samples, buffer provides an efficient way to remove or exchange salts, as well as fractionating molecules of different molecular weights. Supor (PES) microfiltration devices are ideal for clarifying samples with gross particulate, as well as removing proteins and particulate from samples for HPLC analysis of drugs, amino acids, and antibodies.
A built-in deadstop feature prevents samples from spinning to dryness and the unique paddle shape design prevents the creation of a density gradient, preventing proteins from aggregating. Filters are color coded and laser etched for easy identification. The Microsep sample reservoir and filtrate receiver are constructed of polypropylene to reduce non-specific binding of biomolecules.