Macrosep® Advance Centrifugal Devices, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
MAP003C36 MAP003C38 MAP010C38 MAP030C36 MAP030C38 MAP100C36 MAP100C37 MAP100C38 MAPM02C67 MAPM02C68 MAPM45C67 MAPM45C68 MAP003C37 MAP010C36 MAP010C37 MAP030C37 MAP001C36 MAP001C37 MAP001C38
CA89131-972PK 147.1 CAD
CA89131-972 CA89131-976 CA89131-982 CA89131-984 CA89131-988 CA89131-990 CA89131-992 CA89131-994 CA89131-996 CA89131-998 CA89132-000 CA89132-002 CA89131-974 CA89131-978 CA89131-980 CA89131-986 CA89233-880 CA89233-882 CA89233-884
Macrosep® Advance Centrifugal Devices, Cytiva (Formerly Pall Lab)
Filters Centrifugal Filters
These centrifugal devices can quickly concentrate up to 20 ml of biological sample without valuable sample loss.
  • Provides high recoveries, typically >90%
  • Low protein-binding Omega™ membrane and polypropylene housing minimize losses due to non-specific binding
  • Versatile ultrafiltration Omega™ membrane is available in a variety of MWCOs
  • Built-in deadstop prevents spinning to dryness
  • Color-coded and laser etched for easy identification
Macrosep™ Advance centrifugal devices can be used for a wide range of ultrafiltration applications including theMore Product Information