Instatherm® Filtration Apparatus, Ace Glass Incorporated
Supplier: Ace Glass
3704-01 3704-10
71001-492EA 873.6 CAD
71001-492 71001-498
Instatherm® Filtration Apparatus, Ace Glass Incorporated
Filters Bottle Top Filters
This apparatus differs from the standard 47mm filter apparatus (89083-540), as it uses Ace Glass’ proprietary Instatherm® technology to evenly heat the top 400 ml funnel, thus keeping the viscous materials in a flowing liquid state.
  • Fabricated from borosilicate glass
  • Middle adapter is PTFE with a PTFE snap-ring
  • Interchangeable fritted disk is the coarse, 25 to 50 µm size
This system is excellent for filtering oils or thick slurries and using either 47 mm membranes or filter paper disks. Perfect for sample prep for various ASTM petroleum and polymerMore Product Information