Microliter™ and Gastight® Neuros™ Syringes, Hamilton
Supplier: Hamilton
7653-01 7654-01 65457-01 65458-01 65459-01 65460-01 7634-01 7655-01 7656-01 65460-02 65460-05 65460-10 65460-15 65460-20 7658-01 65457-02 65458-02 65459-02 65460-03 65460-06 65460-11 65460-16 65460-21 65460-04
82020-758EA 149.74 CAD
82020-758 82020-760 89218-264 89218-266 89218-268 89218-270 89218-274 89218-276 89218-278 89230-212 89230-214 89230-216 89230-218 89230-220 10192-100 10852-624 10852-626 10852-628 10852-630 10852-632 10852-634 10852-636 10852-638 10852-640
Microliter™ and Gastight® Neuros™ Syringes, Hamilton
Syringes Syringes, Medical
Hamilton offers the most comprehensive selection of standard and custom syringes in the industry
Each is manufactured to achieve the highest level of accuracy and precision possible. Hamilton Neuros™ syringe technology provides unprecedented functionality for controlled animal injections. An industry first, the Neuros™ accurately dispenses 50 nL to 100 μL of liquid through an ultrafine 30, 32, or 33 gauge needle protected by a custom-designed sleeve. Developed specifically for the neurosciences, the Neuros minimizes injection site damage by increasing needle rigidity and delivering aMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable