Pierce™ 96-well Microdialysis Plates, Thermo Scientific
Supplier: Thermo Scientific
88260 88261 88262
CAPI88260EA 1168.43 CAD
CAPI88260 CAPI88261 CAPI88262
Pierce™ 96-well Microdialysis Plates, Thermo Scientific
Microplates Immunology Microplates and Strips
Convenient and rapid dialysis system for 1 to 96 small samples.
  • High yield of proteins, greater than 90% protein recovery after dialysis
  • Rapid dialysis completed in 2 to 4 hours
  • Plate format conforms to the SBS Microplate Standard
  • Protein and DNA desalting/buffer exchange
  • Removal of dyes after protein labeling
  • Enzyme activity/binding assays and drug binding studies
The available plate seals are made of high-quality polypropylene and help protect samples from contamination and evaporation, especially during long-term storage.
Each 96-well Microdialysis Plate comes pre-loaded with twelve 8-dialysis device strips. Each dialysis device strip can be removed from the plate and individual microdialysis devices easily separated. The sample chamber inside each microdialysis device is formed by two low-binding regenerated cellulose membranes that are rated to retain proteins and other macromolecules >10,000 Daltons while allowing removal of buffer salts and contaminants <1000 Daltons. Surface tension and the small distance between the membranes facilitate highly efficient sample recovery using standard laboratory pipettes. Sample dialysis can be efficiently performed in a standard 96 deep-well plate with a minimal amount of buffer while still providing >95% removal of small molecules.
The Thermo Scientific Pierce 96-well Microdialysis Plates are automation-compatible, microplate dialysis devices (3.5 K or 10 K MWCO) for rapidly processing upto 96 samples with volumes 10 to 100 µL, ideal option for high-throughput applications. Individual microdialysis devices can also be used with 2 mL microcentrifuge tubes for processing single samples.
Delivery information: Includes 12 cartridges of eight microdialysis devices in a 96 deep well plate.