Pierce™ HiPPR™ Detergent Removal Resin, Columns and Plates
Supplier: Thermo Scientific
88305 88306 88307
CAPI88305PK 745.7 CAD
CAPI88305 CAPI88306 CAPI88307
Pierce™ HiPPR™ Detergent Removal Resin, Columns and Plates
Cleaning Agents Decontaminants
The HiPPR™ Detergent Removal Spin Column Kit improves mass spectrometry results by efficiently removing detergents from 25 to 200 µL samples with low protein or peptide concentrations.
  • Optimized-removes >95% of detergent from samples with low-concentrations (1 to 100 µg/ml) of proteins or peptides
  • Fast-sample processing takes less than 15 minutes
  • Effective-eliminates detergent-interference in downstream applications like ELISA, isoelectric focusing and mass spectrometry
Optimized for sample concentrations of 1 to 100 µg/ml,the HiPPR (High Protein and Peptide Recovery) Detergent Removal Resin removes >95% of detergents used in protein extraction and biological sample preparation with minimal loss of protein and peptide
The detergents and surfactants used to prepare protein and peptide samples can interfere with analysis by ELISA, isoelectric focusing and mass spectrometry (MS). Removing detergents from peptide samples is especially challenging and critical for MS analysis because even low concentrations of detergents will contaminate instruments and interfere with column binding, elution and peptide ionization. The HiPPR Detergent Removal Resin is ideal for rapid detergent removal from tryptic digests to improve the results of LC-MS/MS and MALDI-MS analysis as to help maintain column and instrument performance over time.
The Detergent Removal Resin is ideal for removing commonly used detergents, including SDS, Triton X-100, NP-40 and CHAPS, at concentrations of 0.5 to 1% and is available in pre-filled spin columns and 96-well filter spin plates for sample volumes up to 100 µl. For other sample sizes, the detergent removal resin slurry is available with empty spin columns that can be used to make custom spin columns for processing sample volumes of 25 to 200 µl.