Pierce™ 1-Step Human In Vitro Protein Expression Kits, Thermo Scientific
Supplier: Thermo Scientific
88881 88882
CAPI88881EA 684.75 CAD
CAPI88881 CAPI88882
Pierce™ 1-Step Human In Vitro Protein Expression Kits, Thermo Scientific
Protein Expression Protein Expression Systems
Thermo Scientific 1-Step Human In Vitro Protein Expression Kits enable the translation and post-transcriptional modification of full-length proteins from mRNA or plasmid templates with yields of up to 100 µg/ml per reaction. Express full-length functional proteins in 90 minutes using the coupled human cell-free protein expression system.
  • Functional - uses the human translational machinery to express active proteins
  • Convenient - perform transcription and translation in a single step
  • High performance - greater yields compared to rabbit reticulocyte in vitro translation
  • Reliable - express proteins that fail in rabbit reticulocyte systems
The Human IVT Kits are unique HeLa cell lysate-based protein expression systems for in vitro translation or coupled transcription/translation reactions.
Requires 30 °C incubator or water bath.
The 1-Step Coupled Human IVT Kit for DNA is a cell-free system using the cellular transcription and translation machinery from a modified HeLa cell line. The procedure is quick and easy and is an effective alternative to other protein expression methods. Simply add an appropriate expression construct to a mixture of HeLa cell lysate, Accessory Protiens, Reaction Mix and incubate at 30 °C for 90 minutes to overnight.
The 1-Step Human IVT Kit for mRNA also uses the human protein translation machinery to produce functional protein. This kit is recommended for translation of mRNA transcripts containing an ECMV IRES element and mRNA stabilizing features designed into the pT7CFE1 expression vector.
Delivery information: Each kit contains HeLa cell lysate (110 or 500 µl), Accessory proteins (25 or 100 µl), Reaction mix (40 or 200 µl), Nuclease-free water (1.5 ml), Expression vector (pT7CFE1-CHis - 10 µg) and a GFP-positive control vector (pCFE-GFP - 10 µg).