Vacuum Trap, with Reservoir, Ace-Thred Inlet/Outlet
Supplier: Ace Glass
8756-44 8756-144
89063-972EA 2154.76 CAD
89063-972 89083-328
Vacuum Trap, with Reservoir, Ace-Thred Inlet/Outlet
Vacuum Traps
Dry ice vacuum trap that features a 1000 mL reservoir flask
Unit has been modified with the addition of No.15 Ace­Threds on the inlet and outlet for use with "Ace-Safe" tubing connectors to make an easy, safe connect/disconnect of tubing. Simply attach 1/2-inch I.D. tubing to the connector and tighten connector in Ace-Thred with bushing to compress silicone o-ring and make leak-tight seal. Chamber body is 125 mm O.D. with 25 mm annular space. Dry ice chamber is 250 mm deep. Overall height is approximately 54.6 cm (21.5 inches). Fabricated from borosilicate glass.
Ordering information: Reservoir flask is available with either 3 mm bore, 1:5 PTFE stopcock or 0-5 PTFE "Easy-Action" valve off bottom for easy draining. Complete item consists of body, two 89052-954 "Ace-Safe" tubing connectors with silicone o-rings, and two 89059-218 bushings.