NS4 Series 1/4" Flow Dry Break, Quick Disconnect Coupling, Colder Products Company
Supplier: Colder Products
NS4D20006 NS4D22004
89066-490PK 583.97 CAD
89066-490 89066-492 89066-494 89066-496 89066-498 89066-500 89066-502 89066-504 89066-506
NS4 Series 1/4" Flow Dry Break, Quick Disconnect Coupling, Colder Products Company
Fittings and Connectors
1/4" flow quick disconnect couplings offer high flow capacity in a compact size
Materials of construction include glass filled polypropylene, 316 stainless steel and EPDM oring seals.
Engineered with an advanced dry break non spill valve design to eliminate drips and mess enhancing operator and equipment safety. Glass filled polypropylebne for added strength under pressure and color coded for visual identfication.
Ordering information: Accessories not included
Order Now
SPECIFICATIONS
Environmentally Preferable