MPX Series 1/2" Flow, Quick Disconnect Coupling for Bio and Pharma, Colder Products Company
Supplier: Colder Products
MPX17803 MPX17839
16001-626PK 11.12 CAD
16001-626 16001-628
MPX Series 1/2" Flow, Quick Disconnect Coupling for Bio and Pharma, Colder Products Company
Fittings and Connectors
These 1/2" flow quick disconnect couplings are manufactured specifically for the bio and pharma markets
Materials of construction include animal free (BSE/TSE) USP class 6 polysulfone and polycarbonate, Buna - n and platinum cured USP class 6 silicone oring seals.
USP Class 6 and animal free (BSE/TSE) materials are appropriate for the Bio, Pharma and Life Sciences with single use dispoability in mind.
Certifications: USP Class VI, BSE/TSE free materials (ADCF)
Ordering information: Accessories not included