Chrome Plated Brass LC Series 1/4" Flow, Quick Disconnect Coupling, Colder Products Company
Supplier: Colder Products
LC12004 LC12006 LC13004 LC13006 LC15004 LC16005 LC16006 LC20006 LC21006 LC23006 LC40004 LC40006 LCD12004 LCD12006 LCD13004 LCD16005 LCD16006 LCD20004 LCD21004 LCD22005 LCD40004 LCD40006 LCD42005 LCD42006
16001-466PK 245.44 CAD
16001-466 16001-468 16001-470 16001-472 16001-474 16001-476 16001-478 16001-480 16001-482 16001-484 16001-486 16001-488 16001-490 16001-492 16001-494 16001-496 16001-498 16001-500 16001-502 16001-504 16001-506 16001-508 16001-510 16001-514 16001-516 16001-518 16001-520 16001-522 16001-524 16001-526 16001-528 16001-530 16001-532 16001-534 16001-536 16001-538 16001-542 16001-544 16001-546 16001-548 16001-550 16001-552 16001-554 16001-556 16001-560 16001-562 16001-564 16001-566 16001-568 16001-570 16001-572 16001-576 16001-578 16001-580 16001-582 16001-584
Chrome Plated Brass LC Series 1/4" Flow, Quick Disconnect Coupling, Colder Products Company
Fittings and Connectors
These 1/4" flow chrome plated brass quick disconnect couplings are built to withstand the most demanding applications and retain their attractive, refletive appearance
Materials of construction includes chrom plated brass, 316 stainless steel, acetal, Buna N oring seals.
For design flexibility these couplings are available in an NSF listed version and can mate with PLC couplings as well.
Ordering information: Accessories not included
Order Now
SPECIFICATIONS
Environmentally Preferable