DrumQuik PRO Series Drum and Tote, Dip Tube and Connection Technology, Colder Products Company
Supplier: Colder Products
DQPRO20G2NPSST DQPRO2056X4202 DQPRO120208206
89066-564PK 825.11 CAD
89066-564 89066-566 89066-568 89066-570 89097-460 89097-462 89097-464 89097-466 89097-468 89097-472
DrumQuik PRO Series Drum and Tote, Dip Tube and Connection Technology, Colder Products Company
Fittings and Connectors
Engineered to provide safe, secure container change outs with a quick connect and disconnect technology and integrated dip tube
Materials of construction include polypropylene or PVDF, HDPE, 316 stainless steel or an all plastic spring, EPDM, FKM or FFKM o-ring seals.
Convert and open dispensing system into a closed dispensing system with the use of any common pump.
Ordering information: Accessories not included