SM Series 1/16" Flow, Miniature Quick Disconnect Coupling, Colder Products Company
Supplier: Colder Products
SMC0112 SMCD01 SMCDM3 SMF0212 SMFDM3 SMFM3 SMFPM0212 SMFPMD01 SMFPMD02 SMFPMDM3 SMFPMM3 SMM0191 SMM0212 SMMM3 SMMP SMMPM0112 SMMPM0212 SMPT0212
16001-990PK 356.01 CAD
16001-990 16001-992 16001-996 16001-998 16002-002 16002-004 16002-006 16002-008 16002-010 16002-012 16002-016 16002-018 16002-022 16002-024 16002-026 16002-028 16002-030 16002-032 16002-034 16002-036 16002-038 16002-040 16002-042 16002-044 16002-046 16002-048 16002-051 16002-052 16002-054 16002-057 16002-058 16002-060 16002-062 16002-064 16002-066 16002-068 16002-070 16002-072 16002-074 16002-076
SM Series 1/16" Flow, Miniature Quick Disconnect Coupling, Colder Products Company
Fittings and Connectors
1/16" flow miniature quick disconnect couplings provide a reliable and secure alternative to luer type fittings
Materials of construction include polypropylene, acetal and chrome plated brass, 316 stainless steel springs and eithe EPDM or Buna N oring seals.
Unique design allows tubingto rotate preventing kinks and accidental disconnections.
Ordering information: Accessories not included