PLC Series 1/4" Flow, Quick Disconnect Coupling, Colder Products Company
Supplier: Colder Products
PLC1000412 PLC1600412 PLC16005 PLC1600612 PLC1700612 PLC21006 PLC2300612 PLC42005 PLCD1700412 PLCD17005 PLCD1700612 PLCD2200612 PLCD40006
16001-640PK 569.04 CAD
16001-640 16001-652 16001-656 16001-658 16001-662 16001-670 16001-676 16001-680 16001-682 16001-686 16001-688 16001-690 16001-692 16001-694 16001-698 16001-702 16001-704 16001-710 16001-716 16001-720 16001-722 16001-728 16001-734 16001-736 16001-738 16001-742 16001-744 16001-752 16001-754 16001-758 16001-764 16001-766 16001-770 16001-772 16001-776 16001-778 16001-780 16001-782 16001-784 16001-788 16001-794 16001-798 16001-800 16001-804
PLC Series 1/4" Flow, Quick Disconnect Coupling, Colder Products Company
Fittings and Connectors
1/4" flow quick disconnect couplings available in either polypropelene or acetal offer good chemical resistance for most applications
Materials of construction include polypropylene or acetal, 316 stainless steel and Buna N or EPDM oring seals.
Availabe with NSF listed materials and can mate with LC seried connectors for design flexibility.
Ordering information: Accessories not included