CQG ChemQuik Series 3/8" Flow, Quick Disconnect Coupling, Colder Products Company
Supplier: Colder Products
CQGD06100106 CQGD06100108 CQGD06100112 CQGD06100208 CQGD06200106 CQGD06200108 CQGD06200112 CQGD06200208
16001-432PK 394.45 CAD
16001-432 16001-434 16001-436 16001-438 16001-440 16001-442 16001-444 16001-446
CQG ChemQuik Series 3/8" Flow, Quick Disconnect Coupling, Colder Products Company
Fittings and Connectors
These 3/8" pressure balanced, non-spill Quick Disconnect Couplings are intended for high purity and aggressive chemical applications
Materials of construction include natural polypropylene, non-wetted Hastelloy C spring, FKM and FFKM o-ring seal options.
A non-metal flow path allows for a braod range of chemical compatibilities.
Ordering information: Accessories not included