Quick Disconnect Coupling, APC Series ¼" Flow, Colder Products Company
Supplier: Colder Products
APC17006SH APCD10004 APCD22004 APCD22006
16001-352PK 128.28 CAD
16001-352 16001-354 16001-356 16001-358 16001-360 16001-362 16001-366 16001-368 16001-370 16001-372 16001-374 16001-376 16001-378 16001-380 16001-382 16001-384 16001-386
Quick Disconnect Coupling, APC Series ¼" Flow, Colder Products Company
Fittings and Connectors
The 1/4" Flow APC series coupling features a plastic thumb latch, fewer moving parts and a smooth contoured design to deliver excelent flow in a compact size
Materials of construction include Acetal, 316 and 302 stainless steel, Buna-N o-ring seal.
The shrouded thumb latch protects against accidental disconnect.
Ordering information: Accessories not included
Order Now
SPECIFICATIONS
Environmentally Preferable