High-Efficiency Condenser, Pilot Plant, Ace-Threds
Supplier: Ace Glass
6016-137 6016-139 6016-141 6016-167 6016-170 6016-176 6016-178 6016-180
89053-958EA 3197.08 CAD
89053-958 89053-960 89053-962 89053-964 89053-966 89053-968 89053-970 89053-972
High-Efficiency Condenser, Pilot Plant, Ace-Threds
Condensers
Highly efficient Pilot Plant Condenser
May be used either for "through" condensation, or for refluxing. Internal baffling acts as impinging surface for entrained particles and discourages diffusion loss. Comes with either standard taper [ST] or spherical [SJ] joints at top and bottom. Fabricated from borosilicate glass. Has No.11 Ace-Thred connections on inlet and outlet, with barbs for 1/4-inch I.D. tubing.