High Capacity, Triple Coil Condenser, Ace Glass Incorporated
Supplier: Ace Glass
6017-210 6017-212
89083-336EA 2852.38 CAD
89083-336 89083-338
High Capacity, Triple Coil Condenser, Ace Glass Incorporated
Condensers
This high capacity, triple coil condenser features a new design that gives even higher throughput than double coil or any rotary evaporator condensers with three internal cooling coils
Designed also for pilot plants and larger reactors where a lot of cooling area is needed for condensing, or for higher temperature reactions where more throughput is needed. Has No.15 Ace-Thred inlet/outlet ports. Top and bottom connections are 2-inch beaded pipe. Overall length is approximately 460 mm and O.D. is approximately 110 mm. Fabricated from borosilicate glass.