SoftGrip™ Multi-Channel Adjustable Volume Manual Pipettes, Hamilton Company
Supplier: Hamilton
55019-50 55019-52 55019-54 55019-56
89050-682EA 1602.48 CAD
89050-682 89050-684 89050-686 89050-688
SoftGrip™ Multi-Channel Adjustable Volume Manual Pipettes, Hamilton Company
Pipettes Micropipettes
The Hamilton line of pipettes puts a priority on addressing the issues of hand strain, fatigue and other injuries.
With the multi-channel pipettes you have the option of either a 5 to 50 µl or 30 to 300 µl in both the 8 and 12 channels. The unique, long tip ejector lever and movable lower body can be rotated a full 360° relative to the handle to provide maximum comfort for left- or right-handed use. The isolated volume adjustment makes it so there's no need for a locking mechanism to prevent accidental volume adjustment.
The new generation of Hamilton pipettes has a dual indicator thatMore Product Information