SoftGrip™ Single-Channel Adjustable Volume Manual Pipettors, Hamilton
Supplier: Hamilton
55019-30 55019-32 55019-34 55019-36 55019-40 55019-38
89050-650EA 677.21 CAD
89050-650 89050-652 89050-654 89050-656 89050-660 89050-658
SoftGrip™ Single-Channel Adjustable Volume Manual Pipettors, Hamilton
Pipettes Micropipettes
The Hamilton line of pipettes puts a priority on addressing the issues of hand strain, fatigue and other injuries.
  • Slim ejection sleeve is made from a high strength, chemically resistant polymer
  • Slim design allows the pipette to reach to the bottom of narrow tubes without the need for extended length tips
SoftGrip™ adjustable volume pipettes are available in 0.2 µl to 1 ml. The isolated volume adjustment makes it so there's no need for a locking mechanism to prevent accidental volume adjustment.
The new generation of Hamilton pipettes has a dualMore Product Information