SoftGrip™ Single-Channel Fixed Volume Manual Pipettors, Hamilton Company
Supplier: Hamilton
55019-01 55019-05 55019-07 55019-09 55019-11 55019-13 55019-15 55019-17 55019-19 55019-03
89050-662EA 400.63 CAD
89050-662 89050-666 89050-668 89050-670 89050-672 89050-674 89050-676 89050-678 89050-680 89221-460
SoftGrip™ Single-Channel Fixed Volume Manual Pipettors, Hamilton Company
Pipettes Micropipettes
The Hamilton line of pipettors puts a priority on addressing the issues of hand strain, fatigue, and other injuries.
  • Slim ejection sleeve is made from a high strength, chemically resistant polymer
  • Slim design allows the pipette to reach to the bottom of narrow tubes without the need for extended length tips
The SoftGrip™ fixed volume pipettors are available in volume ranges from 5 µl to 1ml.
SoftGrip™ pipettors have a universal calibration key that reduces the need for costly calibration services. Calibration requires only minutes to perform and eliminates the downtime and cost of alternative calibration methods.
The SoftGrip™ tip nozzle ensures a proper fit with universal pipettor tips. Unique color coding and labeling clearly indicates the volume of the pipettor.
Hamilton products set new standards for quality, comfort and precision.