Long Path Condenser, Pilot Plant, Ace Glass Incorporated
Supplier: Ace Glass
6012-16 6012-17
89053-904EA 2880.3 CAD
89053-904 89053-906
Long Path Condenser, Pilot Plant, Ace Glass Incorporated
Condensers
This long path condenser creates a turbulent flow, making it very desirable for use under reduced pressure
Internal and external cooling surfaces result in high efficiency per unit length. Has angled standard taper 24/40 reinforced outer joint at top and vertical standard taper 24/40 inner joint at bottom. Fabricated from borosilicate glass. Hose barbs on inlet/outlet are for use with 7/16-inch or 1/2-inch I.D. tubing, size F hose connections.
Order Now
SPECIFICATIONS
Environmentally Preferable