Liebig Condenser, 'No Hold Up', Fully Jacketed, Ace Glass Incorporated
Supplier: Ace Glass
5999-12 5999-14
89053-860EA 569.32 CAD
89053-860 89053-912
Liebig Condenser, 'No Hold Up', Fully Jacketed, Ace Glass Incorporated
Condensers
This Liebig condenser is fully jacketed with water cooled standard taper 24/40 reinforced outer joint at top and standard taper 24/40 inner joint at bottom
Fabricated from borosilicate glass. Has hose barbs for use with 3/8-inch I.D. tubing, size D hose connections.