Liebig Condenser, 'No Hold Up', Ace Glass Incorporated
Supplier: Ace Glass
5997-134 5997-33 5997-34 5997-132 5997-133 5997-32
89053-814EA 515.26 CAD
89053-814 89053-816 89053-818 89053-916 89053-918 89053-922
Liebig Condenser, 'No Hold Up', Ace Glass Incorporated
Condensers
No Hold Up Liebig condenser with standard taper 24/40 reinforced outer joint at top and standard taper 24/40 inner joint at bottom
Fabricated from borosilicate glass. Has #7 Ace-Thred inlet/outlet for connecting "Ace-Safe" hose connections with barb for 1/4-inch I.D. tubing.
Ordering information: Available complete with two 89052-936 "Ace-Safe" connectors with o-rings, or as glassware only.