Graham Condenser, With Tube, Ace Glass Incorporated
Supplier: Ace Glass
5979-12 5979-14
89053-950EA 469.95 CAD
89053-950 89053-952
Graham Condenser, With Tube, Ace Glass Incorporated
Condensers
Graham condenser with standard taper 24/40 reinforced outer joint at top
Has a straight 12mm O.D. tube at bottom for insertion into a rubber stopper. Fabricated from borosilicate glass. For jacket connections, use with 3/8-inch I.D. tubing, size D hose connections.
Order Now
SPECIFICATIONS
Environmentally Preferable