Microliter™ and Gastight® Chaney Adapters, Hamilton Company
Supplier: Hamilton
14725 32146 14700
60374-016EA 298.17 CAD
60374-016 60374-022 60374-024
Microliter™ and Gastight® Chaney Adapters, Hamilton Company
Syringes Syringes for Chromatography
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry
Hamilton Chaney adapters are designed to work with 700, 800, 7000 series Microliter™ syringes and 1700 and 1800 series Gastight® syringes. These adapters are designed to give consistent and reproducible injections and also prevent plungers from bending.
Each is manufactured to achieve the highest level of accuracy and precision possible.